Assay Details
| Gene Symbol: COL17A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000494.3 | NP_000485.3 | 42 - 43 | 2993 | 61 | |
| GenBank mRNA | AB900157.1 | - | 41 - 42 | 2824 | 61 | |
| M91669.1 | - | 42 - 43 | 2929 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001637.hg.1
Set Membership: |
|

COL17A1
Genomic Map