Assay Details
| Gene Symbol: TNR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003285.2 | NP_003276.3 | 20 - 21 | 4183 | 67 | |
| XM_011509949.2 | XP_011508251.1 | 19 - 20 | 3792 | 67 | ||
| XM_017002218.1 | XP_016857707.1 | 21 - 22 | 4620 | 67 | ||
| XM_017002219.1 | XP_016857708.1 | 20 - 21 | 4548 | 67 | ||
| GenBank mRNA | BC129830.1 | - | 18 - 19 | 3506 | 40114168 | 67 |
| X98085.1 | - | 19 - 20 | 3745 | 67 | ||
| Z67996.1 | - | 18 - 19 | 3709 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003548.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
cell adhesion
neuron cell-cell adhesion
axon guidance
associative learning
telencephalon cell migration
negative regulation of cell-cell adhesion
extracellular matrix organization
locomotory exploration behavior
negative regulation of axon extension involved in regeneration
negative regulation of synaptic transmission
synapse organization
neuromuscular process controlling balance
positive regulation of synaptic transmission, glutamatergic
positive regulation of transmission of nerve impulse
long-term synaptic potentiation
cell adhesion
neuron cell-cell adhesion
axon guidance
associative learning
telencephalon cell migration
negative regulation of cell-cell adhesion
extracellular matrix organization
locomotory exploration behavior
negative regulation of axon extension involved in regeneration
negative regulation of synaptic transmission
synapse organization
neuromuscular process controlling balance
positive regulation of synaptic transmission, glutamatergic
positive regulation of transmission of nerve impulse
long-term synaptic potentiation
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01054986.hg.1 | ENST00000263525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054986.hg.1 | ENST00000367673.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054986.hg.1 | ENST00000367674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054986.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054986.hg.1 | TC01003548.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000422274.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000422274.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | ENST00000422274.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054985.hg.1 | ENST00000263525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054985.hg.1 | ENST00000367673.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054985.hg.1 | ENST00000367674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054985.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054985.hg.1 | TC01003548.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000422274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000422274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | ENST00000422274.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018534.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244079.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244079.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244079.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244079.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244079.hg.1 | TC0100018534.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100244080.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244080.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244080.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244080.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244080.hg.1 | TC0100018534.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100244078.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244078.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244078.hg.1 | ENST00000263525.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244078.hg.1 | NM_003285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244078.hg.1 | TC0100018534.hg.1 | Clariom D TC |

TNR
Genomic Map