Assay Details
| Gene Symbol: FBXO44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014765.1 | NP_001014765.1 | 5 - 6 | 988 | 63 | |
| NM_001304791.1 | NP_001291720.1 | 4 - 5 | 727 | 63 | ||
| NM_033182.5 | NP_149438.2 | 4 - 5 | 616 | 63 | ||
| XM_005263535.1 | XP_005263592.1 | 4 - 5 | 558 | 63 | ||
| XM_005263536.4 | XP_005263593.1 | 5 - 6 | 952 | 63 | ||
| XM_005263537.1 | XP_005263594.1 | 4 - 5 | 698 | 63 | ||
| XM_006711043.2 | XP_006711106.1 | 4 - 5 | 558 | 63 | ||
| XM_006711045.2 | XP_006711108.1 | 4 - 5 | 589 | 63 | ||
| XM_011542435.1 | XP_011540737.1 | 5 - 6 | 1020 | 63 | ||
| XM_017002842.1 | XP_016858331.1 | 4 - 5 | 558 | 63 | ||
| XM_017002843.1 | XP_016858332.1 | 4 - 5 | 589 | 63 | ||
| GenBank mRNA | AK293721.1 | - | 4 - 5 | 535 | 63 | |
| AK301418.1 | - | 4 - 5 | 596 | 63 | ||
| AK316284.1 | - | 3 - 4 | 735 | 63 | ||
| AY040878.1 | - | 4 - 5 | 572 | 63 | ||
| BM546141.1 | - | 4 - 5 | 577 | 63 | ||
| BM903606.1 | - | 623 | 63 | |||
| CR749368.1 | - | 5 - 6 | 977 | 63 | ||
More Information
Set Membership: |
|

FBXO44
Genomic Map