Assay Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153260.2 | NP_694992.2 | 2 - 3 | 218 | 77 | |
| XM_011521423.2 | XP_011519725.1 | 1 - 2 | 106 | 77 | ||
| XM_011521424.2 | XP_011519726.1 | 1 - 2 | 106 | 77 | ||
| GenBank mRNA | AK094131.1 | - | 2 - 3 | 212 | 77 | |
| AK094891.1 | - | 1 - 2 | 452 | 77 | ||
| AK309861.1 | - | 2 - 3 | 207 | 77 | ||
| BC058935.1 | - | 2 - 3 | 174 | 5578024 | 77 | |
| BX538324.1 | - | 1 - 2 | 364 | 77 | ||
| CR749223.1 | - | 2 - 3 | 187 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001266.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009518.hg.1 | ENST00000417035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011674.hg.1 | ENST00000323443.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011674.hg.1 | ENST00000397130.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011674.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011674.hg.1 | TC15001266.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011677.hg.1 | ENST00000323443.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011677.hg.1 | ENST00000397130.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011677.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011677.hg.1 | TC15001266.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011673.hg.1 | ENST00000323443.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011673.hg.1 | ENST00000397130.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011673.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011673.hg.1 | TC15001266.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011676.hg.1 | ENST00000323443.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011676.hg.1 | ENST00000397130.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011676.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011676.hg.1 | TC15001266.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | TC1500009192.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166841.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009518.hg.1 | ENST00000417035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | TC1500009192.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166835.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | TC1500009192.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166836.hg.1 | NM_153260.2 | Refseq |

LRRC57
Genomic Map