Assay Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153260.2 | NP_694992.2 | 3 - 4 | 355 | 88 | |
| XM_011521423.2 | XP_011519725.1 | 2 - 3 | 243 | 88 | ||
| XM_011521424.2 | XP_011519726.1 | 2 - 3 | 243 | 88 | ||
| GenBank mRNA | AK094131.1 | - | 3 - 4 | 349 | 88 | |
| AK094891.1 | - | 2 - 3 | 589 | 88 | ||
| BC058935.1 | - | 3 - 4 | 311 | 5578024 | 88 | |
| CR749223.1 | - | 3 - 4 | 324 | 88 | ||
| DA749946.1 | - | 3 - 4 | 355 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001266.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011672.hg.1 | ENST00000323443.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011672.hg.1 | ENST00000397130.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011672.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011672.hg.1 | TC15001266.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000562868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009192.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | NM_153260.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | TC1500009192.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | ENST00000323443.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | ENST00000397130.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | ENST00000563454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | ENST00000569830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | ENST00000570160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166833.hg.1 | NM_153260.2 | Refseq |

LRRC57
Genomic Map