Assay Details
| Gene Symbol: C6orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145020.1 | NP_001138492.1 | 3 - 4 | 392 | 68 | |
| NR_026737.1 | 2 - 3 | 310 | 68 | |||
| XM_006715088.3 | XP_006715151.1 | 3 - 4 | 561 | 68 | ||
| XM_011514572.2 | XP_011512874.1 | 2 - 3 | 357 | 68 | ||
| XM_011514574.2 | XP_011512876.1 | 2 - 3 | 357 | 68 | ||
| XM_011514575.2 | XP_011512877.1 | 3 - 4 | 543 | 68 | ||
| GenBank mRNA | AI809192.1 | - | 2 - 3 | 68 | ||
| AW183566.1 | - | 2 - 3 | 68 | |||
| BC016820.1 | - | 2 - 3 | 206 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001265.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06015159.hg.1 | ENST00000259983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015159.hg.1 | ENST00000379586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015159.hg.1 | ENST00000426700.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015159.hg.1 | ENST00000460742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015159.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015159.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015159.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015159.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015159.hg.1 | TC06001265.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015157.hg.1 | ENST00000259983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015157.hg.1 | ENST00000379586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015157.hg.1 | ENST00000426700.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015157.hg.1 | ENST00000460742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015157.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015157.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015157.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015157.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015157.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015157.hg.1 | TC06001265.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173409.hg.1 | TC0600010790.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173411.hg.1 | TC0600010790.hg.1 | Clariom D TC |

C6orf52
Genomic Map