| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002904.hg.1 |
ENST00000205890.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002904.hg.1 |
NM_016239.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002904.hg.1 |
TC17000205.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700150103.hg.1 |
ENST00000205890.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150103.hg.1 |
ENST00000412324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150103.hg.1 |
ENST00000615845.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150103.hg.1 |
NM_016239.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150103.hg.1 |
TC1700007100.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700150103.hg.1 |
ENST00000205890.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150103.hg.1 |
ENST00000412324.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150103.hg.1 |
ENST00000615845.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150103.hg.1 |
NM_016239.3 |
Refseq |