Assay Details
| Gene Symbol: MYO15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016239.3 | NP_057323.3 | 4 - 5 | 4094 | 75 | |
| XM_011523918.2 | XP_011522220.1 | 3 - 4 | 4454 | 75 | ||
| XM_017024714.1 | XP_016880203.1 | 3 - 4 | 4454 | 75 | ||
| XM_017024715.1 | XP_016880204.1 | 3 - 4 | 4454 | 75 | ||
| GenBank mRNA | AF144094.1 | - | 4 - 5 | 4094 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000205.hg.1
Set Membership: |
|

MYO15A
Genomic Map