| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03025996.hg.1 |
ENST00000535733.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03025996.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03025996.hg.1 |
TC03001440.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011145.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03025994.hg.1 |
ENST00000310914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03025994.hg.1 |
ENST00000535733.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03025994.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03025994.hg.1 |
TC03001440.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03025995.hg.1 |
ENST00000310914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03025995.hg.1 |
ENST00000535733.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03025995.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03025995.hg.1 |
TC03001440.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011145.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183057.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183057.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183057.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183057.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183057.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183057.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183057.hg.1 |
TC0300011145.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300183058.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183058.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183058.hg.1 |
TC0300011145.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300183055.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183055.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183055.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183055.hg.1 |
ENST00000310914.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183055.hg.1 |
ENST00000314534.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183055.hg.1 |
NM_152397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183055.hg.1 |
TC0300011145.hg.1 |
Clariom D TC |