Assay Details
| Gene Symbol: C6orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145020.1 | NP_001138492.1 | 2 - 3 | 194 | 95 | |
| XM_006715088.3 | XP_006715151.1 | 2 - 3 | 363 | 95 | ||
| GenBank mRNA | AI809192.1 | - | 3 - 4 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001265.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06015160.hg.1 | ENST00000259983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015160.hg.1 | ENST00000426700.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015160.hg.1 | ENST00000460742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015160.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015160.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015160.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015160.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015160.hg.1 | TC06001265.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015161.hg.1 | ENST00000259983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015161.hg.1 | ENST00000426700.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015161.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015161.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015161.hg.1 | TC06001265.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173413.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173413.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173413.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173413.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173413.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173413.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173413.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173413.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173413.hg.1 | TC0600010790.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000379586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000467832.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010790.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | ENST00000259983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | ENST00000426700.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | ENST00000460742.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | ENST00000503680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | NM_001145020.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | NR_026736.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | NR_026737.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600173412.hg.1 | TC0600010790.hg.1 | Clariom D TC |

C6orf52
Genomic Map