| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012480.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012480.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012480.hg.1 |
TC0X000929.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012481.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012481.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X012481.hg.1 |
TC0X000929.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00163665.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163665.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163665.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163665.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163665.hg.1 |
TC0X00009271.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00163666.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163666.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163666.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00163666.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00163666.hg.1 |
TC0X00009271.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
ENST00000379044.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009271.hg.1 |
NM_139058.2 |
Refseq |