Assay Details
| Gene Symbol: ABCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288985.1 | NP_001275914.1 | 24 - 25 | 3487 | 65 | |
| NM_001288986.1 | NP_001275915.1 | 23 - 24 | 3326 | 65 | ||
| NM_007168.3 | NP_009099.1 | 22 - 23 | 3206 | 65 | ||
| XM_005256938.2 | XP_005256995.1 | 23 - 24 | 3354 | 65 | ||
| XM_005256940.4 | XP_005256997.1 | 22 - 23 | 3259 | 65 | ||
| XM_011524191.2 | XP_011522493.1 | 23 - 24 | 3317 | 65 | ||
| GenBank mRNA | AB020629.1 | - | 22 - 23 | 3165 | 65 | |
| AK304286.1 | - | 22 - 23 | 3158 | 65 | ||
| BC130280.1 | - | 24 - 25 | 3336 | 40146311 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001825.hg.1
Set Membership: |
|

ABCA8
Genomic Map