| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023231.hg.1 |
ENST00000311277.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023231.hg.1 |
ENST00000515654.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023231.hg.1 |
NM_001039580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023231.hg.1 |
TC04001667.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400183804.hg.1 |
ENST00000311277.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183804.hg.1 |
ENST00000424373.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183804.hg.1 |
ENST00000433024.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183804.hg.1 |
ENST00000515654.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183804.hg.1 |
NM_001039580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400183804.hg.1 |
ENST00000311277.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183804.hg.1 |
ENST00000424373.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183804.hg.1 |
ENST00000433024.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183804.hg.1 |
ENST00000515654.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183804.hg.1 |
NM_001039580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400183804.hg.1 |
TC0400012193.hg.1 |
Clariom D TC |