| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003391.hg.1 |
NM_001172713.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003391.hg.1 |
NM_002078.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003391.hg.1 |
TC03000178.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003387.hg.1 |
ENST00000356847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003387.hg.1 |
ENST00000361924.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003387.hg.1 |
ENST00000444882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003387.hg.1 |
NM_001172713.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003387.hg.1 |
NM_002078.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003387.hg.1 |
TC03000178.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300149149.hg.1 |
NM_001172713.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149149.hg.1 |
NM_002078.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149149.hg.1 |
NM_001172713.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149149.hg.1 |
NM_002078.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149149.hg.1 |
TC0300007044.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
ENST00000356847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
ENST00000361924.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
ENST00000437131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
ENST00000444882.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
NM_001172713.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
NM_002078.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
ENST00000356847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
ENST00000361924.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
ENST00000437131.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
ENST00000444882.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
NM_001172713.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
NM_002078.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149145.hg.1 |
TC0300007044.hg.1 |
Clariom D TC |