| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022049.hg.1 |
ENST00000336686.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022049.hg.1 |
ENST00000354379.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022049.hg.1 |
ENST00000396428.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022049.hg.1 |
ENST00000421276.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022049.hg.1 |
ENST00000421307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022049.hg.1 |
ENST00000440230.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022049.hg.1 |
NM_001134369.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022049.hg.1 |
NM_006309.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022049.hg.1 |
NM_017724.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022049.hg.1 |
TC03001286.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000336686.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000354379.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000396428.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000421276.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000440230.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000460646.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000496479.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
NM_001134369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
NM_001282691.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
NM_006309.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
NM_017724.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000336686.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000354379.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000396428.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000421276.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000440230.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000460646.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
ENST00000496479.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
NM_001134369.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
NM_001282691.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
NM_006309.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
NM_017724.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300177998.hg.1 |
TC0300010722.hg.1 |
Clariom D TC |