| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR0300178028.hg.1 |
ENST00000336686.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178028.hg.1 |
ENST00000396428.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178028.hg.1 |
ENST00000461672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178028.hg.1 |
NM_001282691.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178028.hg.1 |
NM_006309.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178028.hg.1 |
ENST00000336686.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178028.hg.1 |
ENST00000396428.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178028.hg.1 |
ENST00000461672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300178028.hg.1 |
NM_001282691.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178028.hg.1 |
NM_006309.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300178028.hg.1 |
TC0300010722.hg.1 |
Clariom D TC |