| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015181.hg.1 |
ENST00000439282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
ENST00000237889.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
ENST00000433898.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
ENST00000450023.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
ENST00000454214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
NM_001257102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
NM_002491.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
ENST00000237889.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
ENST00000433898.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
ENST00000450023.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
ENST00000454214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
NM_001257102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
NM_002491.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200173782.hg.1 |
TC0200010438.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100015181.hg.1 |
ENST00000439282.1 |
Ensembl |