Assay Details
| Gene Symbol: SDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144952.1 | NP_001138424.1 | 15 - 16 | 2043 | 86 | |
| XM_011524914.2 | XP_011523216.1 | 15 - 16 | 2427 | 86 | ||
| XM_011524915.2 | XP_011523217.1 | 15 - 16 | 2429 | 86 | ||
| XM_011524916.2 | XP_011523218.1 | 15 - 16 | 2437 | 86 | ||
| GenBank mRNA | AB040947.1 | - | 8 - 9 | 1074 | 86 | |
| BC066363.1 | - | 7 - 8 | 943 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001847.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000585283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000585283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |

SDK2
Genomic Map