Assay Details
| Gene Symbol: SDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144952.1 | NP_001138424.1 | 28 - 29 | 3998 | 118 | |
| XM_011524914.2 | XP_011523216.1 | 28 - 29 | 4382 | 118 | ||
| XM_011524915.2 | XP_011523217.1 | 28 - 29 | 4384 | 118 | ||
| GenBank mRNA | AB040947.1 | - | 21 - 22 | 3029 | 118 | |
| AL137357.1 | - | 6 - 7 | 906 | 118 | ||
| BC045763.1 | - | 10 - 11 | 1526 | 118 | ||
| BC066363.1 | - | 20 - 21 | 2898 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001847.hg.1
Set Membership: |
|

SDK2
Genomic Map