Assay Details
| Gene Symbol: SDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144952.1 | NP_001138424.1 | 36 - 37 | 4982 | 86 | |
| XM_011524914.2 | XP_011523216.1 | 35 - 36 | 5309 | 86 | ||
| XM_011524915.2 | XP_011523217.1 | 36 - 37 | 5368 | 86 | ||
| GenBank mRNA | AB040947.1 | - | 29 - 30 | 4013 | 86 | |
| AK001694.1 | - | 3 - 4 | 369 | 86 | ||
| AL137357.1 | - | 14 - 15 | 1890 | 86 | ||
| BC045763.1 | - | 17 - 18 | 2453 | 86 | ||
| BC066363.1 | - | 28 - 29 | 3882 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001847.hg.1
Set Membership: |
|

SDK2
Genomic Map