Assay Details
| Gene Symbol: SDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144952.1 | NP_001138424.1 | 43 - 44 | 6033 | 103 | |
| XM_011524914.2 | XP_011523216.1 | 42 - 43 | 6360 | 103 | ||
| XM_011524915.2 | XP_011523217.1 | 43 - 44 | 6419 | 103 | ||
| GenBank mRNA | AK001694.1 | - | 10 - 11 | 1420 | 103 | |
| AK091827.1 | - | 3 - 4 | 1031 | 103 | ||
| AL137357.1 | - | 21 - 22 | 2941 | 103 | ||
| BC045763.1 | - | 24 - 25 | 3504 | 103 | ||
| BC066363.1 | - | 35 - 36 | 4933 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001847.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000392650.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000410094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000424778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000479356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000585283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | ENST00000585283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011653.hg.1 | NM_001144952.1 | Refseq |

SDK2
Genomic Map