Assay Details
| Gene Symbol: SYNPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007286.5 | NP_009217.3 | 2 - 3 | 2573 | 107 | |
| XM_005268370.1 | XP_005268427.1 | 2 - 3 | 2516 | 107 | ||
| XM_005268371.1 | XP_005268428.1 | 2 - 3 | 2436 | 107 | ||
| XM_006714755.3 | XP_006714818.1 | 3 - 4 | 3125 | 107 | ||
| XM_017009007.1 | XP_016864496.1 | 3 - 4 | 3158 | 107 | ||
| XM_017009008.1 | XP_016864497.1 | 3 - 4 | 3097 | 107 | ||
| GenBank mRNA | AB028952.1 | - | 2 - 3 | 2527 | 107 | |
| AF499136.1 | - | 2 - 3 | 2487 | 107 | ||
| AF499137.1 | - | 2 - 3 | 2487 | 107 | ||
| BC152538.1 | - | 2 - 3 | 2527 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000833.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05012010.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012010.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012010.hg.1 | TC05000833.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000518872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000518872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000518872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000518872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163590.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163590.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163590.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163590.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163590.hg.1 | TC0500009097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500163591.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163591.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163591.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163591.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163591.hg.1 | TC0500009097.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 140667 | - | - |

SYNPO
Genomic Map