Assay Details
| Gene Symbol: RNPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286625.1 | NP_001273554.1 | 3 - 4 | 503 | 62 | |
| NM_001286626.1 | NP_001273555.1 | 2 - 3 | 510 | 62 | ||
| NM_001286627.1 | NP_001273556.1 | 2 - 3 | 510 | 62 | ||
| NM_006711.4 | NP_006702.1 | 3 - 4 | 564 | 62 | ||
| NM_080594.3 | NP_542161.1 | 3 - 4 | 698 | 62 | ||
| NR_104485.1 | 2 - 3 | 632 | 62 | |||
| XM_005255048.1 | XP_005255105.1 | 3 - 4 | 791 | 62 | ||
| XM_005255049.3 | XP_005255106.1 | 3 - 4 | 2256 | 62 | ||
| XM_017022874.1 | XP_016878363.1 | 3 - 4 | 1157 | 62 | ||
| GenBank mRNA | AB445101.1 | - | 2 - 3 | 223 | 62 | |
| AF015608.1 | - | 2 - 3 | 237 | 62 | ||
| AF274003.1 | - | 2 - 3 | 199 | 62 | ||
| AK289955.1 | - | 4 - 5 | 458 | 62 | ||
| AK293343.1 | - | 2 - 3 | 215 | 62 | ||
| AK316132.1 | - | 2 - 3 | 500 | 62 | ||
| BC001659.2 | - | 3 - 4 | 388 | 2959954 | 62 | |
| BC001838.2 | - | 3 - 4 | 388 | 2959954 | 62 | |
| BC108316.1 | - | 3 - 4 | 509 | 6151864 | 62 | |
| BG716139.1 | - | 3 - 4 | 406 | 62 | ||
| BI819738.1 | - | 2 - 3 | 193 | 62 | ||
| BQ059026.1 | - | 2 | 366 | 62 | ||
| DA728900.1 | - | 3 - 4 | 400 | 62 | ||
| L37368.1 | - | 3 - 4 | 772 | 62 | ||
More Information
Set Membership: |
|

RNPS1
Genomic Map