Assay Details
| Gene Symbol: NDUFV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021074.4 | NP_066552.2 | 1 - 2 | 165 | 117 | |
| GenBank mRNA | AK291865.1 | - | 1 - 2 | 61 | 117 | |
| AK312129.1 | - | 1 - 2 | 83 | 117 | ||
| BC001632.1 | - | 1 - 2 | 75 | 3536345 | 117 | |
| BC017487.1 | - | 1 - 2 | 73 | 3537815 | 117 | |
| BG742948.1 | - | 1 - 2 | 165 | 117 | ||
| CB121843.1 | - | 1 - 2 | 118 | 117 | ||
| M22538.1 | - | 1 - 2 | 69 | 117 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000041.hg
Additional Information:
This assay is in the 5'UTR region of the AK291865.1,AK312129.1,BC001632.1,BC017487.1,M22538.1,NM_021074.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial electron transport, NADH to ubiquinone
nervous system development
mitochondrial respiratory chain complex I assembly
cardiac muscle tissue development
NADH dehydrogenase (ubiquinone) activity
electron carrier activity
metal ion binding
2 iron, 2 sulfur cluster binding
mitochondrial electron transport, NADH to ubiquinone
nervous system development
mitochondrial respiratory chain complex I assembly
cardiac muscle tissue development
NADH dehydrogenase (ubiquinone) activity
electron carrier activity
metal ion binding
2 iron, 2 sulfur cluster binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18000419.hg.1 | ENST00000318388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000419.hg.1 | ENST00000497577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000419.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000419.hg.1 | TC18000041.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000426.hg.1 | ENST00000318388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000426.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000426.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000426.hg.1 | TC18000041.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000583375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000583375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000415.hg.1 | ENST00000318388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000415.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000415.hg.1 | TC18000041.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000583375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | TC1800009214.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | ENST00000583375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163195.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000583375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000583375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163179.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163179.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163179.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163179.hg.1 | TC1800009214.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800163179.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163179.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163179.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163184.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163184.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163184.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163184.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163184.hg.1 | TC1800009214.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800163184.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163184.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163184.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163184.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163180.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163180.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163180.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163180.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163180.hg.1 | TC1800009214.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800163180.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163180.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163180.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163180.hg.1 | NM_021074.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 155598 | - | - |

NDUFV2
Genomic Map