| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18000427.hg.1 |
ENST00000318388.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18000427.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18000427.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18000427.hg.1 |
TC18000041.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000474740.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000474740.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000474740.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000497577.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000497577.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000497577.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000497577.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000583375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
ENST00000583375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000465096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000474740.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000474740.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000474740.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000497577.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000497577.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000497577.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000497577.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000577703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000583375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
ENST00000583375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009214.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800163197.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163197.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163197.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163197.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163197.hg.1 |
NM_021074.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800163197.hg.1 |
TC1800009214.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800163197.hg.1 |
ENST00000318388.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163197.hg.1 |
ENST00000400033.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163197.hg.1 |
ENST00000474350.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163197.hg.1 |
ENST00000483511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163197.hg.1 |
NM_021074.4 |
Refseq |