Assay Details
| Gene Symbol: TMEM59L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012109.2 | NP_036241.1 | 4 - 5 | 652 | 131 | |
| GenBank mRNA | AF186264.1 | - | 4 - 5 | 622 | 131 | |
| AK309384.1 | - | 4 - 5 | 651 | 131 | ||
| BC010446.1 | - | 4 - 5 | 593 | 4181442 | 131 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000362.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19004791.hg.1 | ENST00000262817.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004791.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004791.hg.1 | TC19000362.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | ENST00000594709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | TC1900007399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | ENST00000594709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900154388.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000262817.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000594859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000598660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | ENST00000600490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007399.hg.1 | NM_012109.2 | Refseq |

TMEM59L
Genomic Map