Assay Details
| Gene Symbol: LRRFIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137552.1 | NP_001131024.1 | 2 - 3 | 444 | 86 | |
| NM_004735.3 | NP_004726.2 | 2 - 3 | 444 | 86 | ||
| GenBank mRNA | AJ223075.1 | - | 2 - 3 | 330 | 86 | |
| AK307485.1 | - | 2 - 3 | 246 | 86 | ||
| BC108914.1 | - | 2 - 3 | 269 | 40027218 | 86 | |
More Information
Set Membership: |
|

LRRFIP1
Genomic Map