| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026017.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026017.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026017.hg.1 |
TC03001442.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026016.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026016.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026016.hg.1 |
TC03001442.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026018.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026018.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026018.hg.1 |
TC03001442.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026019.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026019.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026019.hg.1 |
TC03001442.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183074.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183074.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183074.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183074.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183074.hg.1 |
TC0300011146.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300183078.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183078.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183078.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183078.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183078.hg.1 |
TC0300011146.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300183075.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183075.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183075.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183075.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183075.hg.1 |
TC0300011146.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300183076.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183076.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183076.hg.1 |
ENST00000232888.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300183076.hg.1 |
NM_004704.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300183076.hg.1 |
TC0300011146.hg.1 |
Clariom D TC |