Assay Details
| Gene Symbol: APEX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014481.3 | NP_055296.2 | 2 - 3 | 345 | 126 | |
| GenBank mRNA | AB021260.1 | - | 2 - 3 | 305 | 126 | |
| AB049211.1 | - | 2 - 3 | 268 | 126 | ||
| AF119046.1 | - | 2 - 3 | 311 | 126 | ||
| AJ011311.1 | - | 2 - 3 | 311 | 126 | ||
| BC002959.1 | - | 2 - 3 | 283 | 3537317 | 126 | |
| BI837686.1 | - | 2 - 3 | 321 | 126 | ||
| BP363514.1 | - | 2 - 3 | 345 | 126 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000326.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004348.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004348.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004348.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004348.hg.1 | TC0X000326.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004346.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004346.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004346.hg.1 | TC0X000326.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004349.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004349.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004349.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004349.hg.1 | TC0X000326.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004350.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004350.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004350.hg.1 | TC0X000326.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004351.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004351.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004351.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004351.hg.1 | TC0X000326.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00150651.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150651.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150651.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150651.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150651.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150651.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150651.hg.1 | TC0X00007369.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00150654.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150654.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150654.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150654.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150654.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150654.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150654.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150654.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150654.hg.1 | TC0X00007369.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00150655.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150655.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150655.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150655.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150655.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150655.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150655.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150655.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150655.hg.1 | TC0X00007369.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00150656.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150656.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150656.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150656.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150656.hg.1 | TC0X00007369.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007369.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150650.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150650.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150650.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150650.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150650.hg.1 | TC0X00007369.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00150653.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150653.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150653.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150653.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150653.hg.1 | ENST00000374987.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150653.hg.1 | ENST00000471758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00150653.hg.1 | NM_001271748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150653.hg.1 | NM_014481.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00150653.hg.1 | TC0X00007369.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 209621 | - | - |

APEX2
Genomic Map