| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004352.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004352.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004352.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X004352.hg.1 |
TC0X000326.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007369.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150658.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150658.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150658.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150658.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150658.hg.1 |
ENST00000374987.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150658.hg.1 |
ENST00000471758.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00150658.hg.1 |
NM_001271748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150658.hg.1 |
NM_014481.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00150658.hg.1 |
TC0X00007369.hg.1 |
Clariom D TC |