Assay Details
| Gene Symbol: TFE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282142.1 | NP_001269071.1 | 4 - 5 | 1012 | 63 | |
| NM_006521.5 | NP_006512.2 | 4 - 5 | 1077 | 63 | ||
| GenBank mRNA | AK295497.1 | - | 3 - 4 | 568 | 63 | |
| AK313540.1 | - | 4 - 5 | 798 | 63 | ||
| AL161985.1 | - | 4 - 5 | 913 | 63 | ||
| BC026027.1 | - | 4 - 5 | 1024 | 4576858 | 63 | |
| X51330.1 | - | 1 - 2 | 165 | 63 | ||
| X96717.1 | - | 4 - 5 | 1020 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001025.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
transcription from RNA polymerase II promoter
humoral immune response
regulation of osteoclast differentiation
positive regulation of cell adhesion
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription regulatory region DNA binding
protein dimerization activity
adaptive immune response
transcription from RNA polymerase II promoter
humoral immune response
regulation of osteoclast differentiation
positive regulation of cell adhesion
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription regulatory region DNA binding
protein dimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000481606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000481606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000481606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000495940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000495940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | ENST00000495940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013652.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013652.hg.1 | ENST00000493583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013652.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013652.hg.1 | TC0X001025.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013649.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013649.hg.1 | ENST00000493583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013649.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013649.hg.1 | TC0X001025.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013651.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013651.hg.1 | ENST00000493583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013651.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013651.hg.1 | TC0X001025.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000481606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000481606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000481606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000495940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000495940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | ENST00000495940.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009648.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | ENST00000481606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | ENST00000481606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165799.hg.1 | TC0X00009648.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | ENST00000481606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | ENST00000315869.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | ENST00000481606.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | ENST00000487451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | ENST00000493583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | NM_001282142.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | NM_006521.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165800.hg.1 | TC0X00009648.hg.1 | Clariom D TC |

TFE3
Genomic Map