| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023906.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023906.hg.1 |
ENST00000353903.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023906.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023906.hg.1 |
TC11001988.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100187475.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100187475.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100187475.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100187475.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100187475.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100187475.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100187475.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100187475.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100187475.hg.1 |
TC1100011339.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000294288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000353903.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
ENST00000545205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011339.hg.1 |
NM_016366.2 |
Refseq |