Assay Details
| Gene Symbol: ALDH9A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000696.3 | NP_000687.3 | 1 - 2 | 287 | 106 | |
| GenBank mRNA | AF172093.1 | - | 1 - 2 | 110 | 106 | |
| AK312751.1 | - | 1 - 2 | 216 | 106 | ||
| BC070030.1 | - | 1 - 2 | 216 | 106 | ||
| BC151140.1 | - | 1 - 2 | 275 | 9057040 | 106 | |
| BC151141.1 | - | 1 - 2 | 275 | 9057041 | 106 | |
| U34252.1 | - | 1 - 2 | 484 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003466.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
liver development
cellular aldehyde metabolic process
neurotransmitter biosynthetic process
hormone metabolic process
carnitine biosynthetic process
oxidation-reduction process
3-chloroallyl aldehyde dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
aminobutyraldehyde dehydrogenase activity
1-pyrroline dehydrogenase activity
protein homodimerization activity
amine binding
4-trimethylammoniobutyraldehyde dehydrogenase activity
NAD binding
kidney development
liver development
cellular aldehyde metabolic process
neurotransmitter biosynthetic process
hormone metabolic process
carnitine biosynthetic process
oxidation-reduction process
3-chloroallyl aldehyde dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
aminobutyraldehyde dehydrogenase activity
1-pyrroline dehydrogenase activity
protein homodimerization activity
amine binding
4-trimethylammoniobutyraldehyde dehydrogenase activity
NAD binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01053982.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053982.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053982.hg.1 | TC01003466.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053977.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053977.hg.1 | ENST00000461664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053977.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053977.hg.1 | TC01003466.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053980.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053980.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053980.hg.1 | TC01003466.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053974.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053974.hg.1 | ENST00000461664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053974.hg.1 | ENST00000538148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053974.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053974.hg.1 | TC01003466.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053978.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053978.hg.1 | ENST00000461664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053978.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053978.hg.1 | TC01003466.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000384263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000463610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000463610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000463610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000463610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000491436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000491436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000491436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000491436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | ENST00000491436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053979.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053979.hg.1 | ENST00000461664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053979.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01053979.hg.1 | TC01003466.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000384263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000463610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000463610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000463610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000463610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000491436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000491436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000491436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000491436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | ENST00000491436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016260.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223790.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223790.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223790.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223790.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223790.hg.1 | TC0100016260.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100223785.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223785.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223785.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223785.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223785.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223785.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223785.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223785.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223785.hg.1 | TC0100016260.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100223787.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223787.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223787.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223787.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223787.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223787.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223787.hg.1 | ENST00000471457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223787.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223787.hg.1 | TC0100016260.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100223788.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223788.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223788.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223788.hg.1 | ENST00000354775.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223788.hg.1 | ENST00000461664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100223788.hg.1 | NM_000696.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100223788.hg.1 | TC0100016260.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 189412 | - | - |

ALDH9A1
Genomic Map