Assay Details
| Gene Symbol: MAP4K5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006575.4 | NP_006566.2 | 20 - 21 | 1800 | 121 | |
| NM_198794.2 | NP_942089.1 | 20 - 21 | 1848 | 121 | ||
| XM_006720013.3 | XP_006720076.1 | 20 - 21 | 1695 | 121 | ||
| XM_011536377.2 | XP_011534679.1 | 20 - 21 | 1709 | 121 | ||
| XM_011536378.2 | XP_011534680.1 | 20 - 21 | 1545 | 121 | ||
| XM_011536379.2 | XP_011534681.1 | 16 - 17 | 1656 | 121 | ||
| XM_011536380.2 | XP_011534682.1 | 7 - 8 | 2442 | 121 | ||
| GenBank mRNA | AK124738.1 | - | 6 - 7 | 2233 | 121 | |
| AK313612.1 | - | 19 - 20 | 1606 | 121 | ||
| BC036013.1 | - | 20 - 21 | 1738 | 5272190 | 121 | |
| U77129.1 | - | 20 - 21 | 1800 | 121 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001120.hg.1
Set Membership: |
|

MAP4K5
Genomic Map