Assay Details
| Gene Symbol: PRMT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256536.1 | NP_001243465.1 | 4 - 5 | 497 | 103 | |
| NM_019854.4 | NP_062828.3 | 4 - 5 | 906 | 103 | ||
| GenBank mRNA | AF263539.1 | - | 3 - 4 | 371 | 103 | |
| AK315619.1 | - | 4 - 5 | 498 | 103 | ||
| BC022458.1 | - | 4 - 5 | 513 | 4796524 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
histone methylation
peptidyl-arginine methylation
peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
histone arginine methylation
regulation of protein binding
protein binding
histone-arginine N-methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
protein-arginine omega-N monomethyltransferase activity
protein-arginine omega-N asymmetric methyltransferase activity
identical protein binding
protein homodimerization activity
protein heterodimerization activity
regulation of transcription, DNA-templated
histone methylation
peptidyl-arginine methylation
peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
histone arginine methylation
regulation of protein binding
protein binding
histone-arginine N-methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
protein-arginine omega-N monomethyltransferase activity
protein-arginine omega-N asymmetric methyltransferase activity
identical protein binding
protein homodimerization activity
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200144963.hg.1 | TC1200006532.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000261252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000382622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000452611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | ENST00000543701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_001256536.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006532.hg.1 | NM_019854.4 | Refseq |

PRMT8
Genomic Map