Assay Details
| Gene Symbol: CABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300896.1 | NP_001287825.1 | 2 - 3 | 315 | 67 | |
| XM_011545183.2 | XP_011543485.1 | 2 - 3 | 167 | 67 | ||
| XM_017018025.1 | XP_016873514.1 | 2 - 3 | 653 | 67 | ||
| GenBank mRNA | BC033167.1 | - | 2 - 3 | 315 | 4549347 | 67 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC033167.1, NM_001300896.1, XM_011545183.2, XM_017018025.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000602912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000602912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159952.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159952.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159952.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159952.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159952.hg.1 | ENST00000438189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159952.hg.1 | ENST00000545777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159952.hg.1 | NM_001300895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159952.hg.1 | NM_001300896.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159952.hg.1 | TC1100008138.hg.1 | Clariom D TC |

CABP4
Genomic Map