Assay Details
| Gene Symbol: MTSS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005256236.4 | XP_005256293.2 | 6 - 7 | 868 | 76 | |
| XM_011523454.2 | XP_011521756.2 | 6 - 7 | 868 | 76 | ||
| GenBank mRNA | BC002770.1 | - | 3 - 4 | 217 | 76 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000338779.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000562883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000576338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | ENST00000616026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011560.hg.1 | NM_138383.2 | Refseq |

MTSS1L
Genomic Map