Assay Details
| Gene Symbol: USP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017415.1 | NP_001017415.1 | 7 - 8 | 1754 | 73 | |
| NM_001017416.1 | NP_001017416.1 | 7 - 8 | 1659 | 73 | ||
| NM_003368.4 | NP_003359.3 | 7 - 8 | 2184 | 73 | ||
| GenBank mRNA | AB014458.1 | - | 7 - 8 | 1535 | 73 | |
| AB208893.1 | - | 7 - 8 | 1815 | 73 | ||
| AF117386.1 | - | 7 - 8 | 1671 | 73 | ||
| AK312423.1 | - | 7 - 8 | 1847 | 73 | ||
| AL117575.1 | - | 6 - 7 | 1506 | 73 | ||
| BC023216.1 | - | 3 - 4 | 1019 | 73 | ||
| BC050525.1 | - | 7 - 8 | 1888 | 6473568 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000707.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
regulation of DNA repair
ubiquitin-dependent protein catabolic process
response to UV
protein deubiquitination
monoubiquitinated protein deubiquitination
interstrand cross-link repair
DNA damage response, detection of DNA damage
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
skeletal system development
regulation of DNA repair
ubiquitin-dependent protein catabolic process
response to UV
protein deubiquitination
monoubiquitinated protein deubiquitination
interstrand cross-link repair
DNA damage response, detection of DNA damage
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000442679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000442679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000442679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000442679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011323.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011323.hg.1 | ENST00000371146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011323.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011323.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011323.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011323.hg.1 | TC01000707.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000442679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000442679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000442679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000442679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | ENST00000452143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008554.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160740.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160740.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160740.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160740.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160740.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160740.hg.1 | ENST00000339950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160740.hg.1 | ENST00000371146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160740.hg.1 | NM_001017415.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160740.hg.1 | NM_001017416.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160740.hg.1 | NM_003368.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160740.hg.1 | TC0100008554.hg.1 | Clariom D TC |

USP1
Genomic Map