Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | OTU deubiquitinase 4 | OTUD4 |
DUBA6, HIN1, HSHIN1 |
-301.0kb | Anti-sense | Chr. 4 - 145133650 - 145180019 on NCBI Build GRCh38 |
| Downstream | SMAD1 antisense RNA 2 | SMAD1-AS2 | 15.5kb | Anti-sense | Chr. 4 - 145497049 - 145502980 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map