Assay Details
| Gene Symbol: MOB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201403.2 | NP_958805.1 | 1 - 2 | 179 | 121 | |
| GenBank mRNA | BI601393.1 | - | 1 - 2 | 134 | 121 | |
| CF125162.1 | - | 1 - 2 | 78 | 121 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006354.hg
Additional Information:
This assay is in the 5'UTR region of the NM_201403.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01040989.hg.1 | ENST00000319928.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040989.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040989.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040989.hg.1 | TC01006354.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040982.hg.1 | ENST00000271139.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040982.hg.1 | ENST00000319928.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040982.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040982.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040982.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040982.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040982.hg.1 | TC01006354.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040988.hg.1 | ENST00000319928.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040988.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040988.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040988.hg.1 | TC01006354.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100242004.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242004.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242004.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242004.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242004.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242004.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242004.hg.1 | TC0100018438.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018438.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241997.hg.1 | TC0100018438.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | ENST00000271139.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | ENST00000319928.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | ENST00000371940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | ENST00000477318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | NM_145279.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | NM_201403.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241996.hg.1 | TC0100018438.hg.1 | Clariom D TC |

MOB3C
Genomic Map