Assay Details
| Gene Symbol: STAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017564.9 | NP_060034.9 | 47 - 48 | 5120 | 90 | |
| XM_011538537.2 | XP_011536839.1 | 47 - 48 | 5161 | 90 | ||
| XM_011538538.2 | XP_011536840.1 | 47 - 48 | 5161 | 90 | ||
| XM_011538539.2 | XP_011536841.1 | 33 - 34 | 3364 | 90 | ||
| XM_011538541.2 | XP_011536843.1 | 25 - 26 | 2471 | 90 | ||
| XM_011538542.2 | XP_011536844.1 | 14 - 15 | 1280 | 90 | ||
| XM_017019585.1 | XP_016875074.1 | 47 - 48 | 5161 | 90 | ||
| GenBank mRNA | AB052958.1 | - | 47 - 48 | 5116 | 90 | |
| AF160476.1 | - | 2 - 3 | 142 | 90 | ||
| AJ295695.1 | - | 47 - 48 | 5120 | 90 | ||
| AK024503.1 | - | 15 - 16 | 1462 | 90 | ||
| AK074051.1 | - | 25 - 26 | 2491 | 90 | ||
| AK160380.1 | - | 46 - 47 | 4838 | 90 | ||
| AL133021.1 | - | 7 - 8 | 602 | 90 | ||
| AY227444.1 | - | 16 - 17 | 1529 | 90 | ||
| AY311388.1 | - | 47 - 48 | 5138 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000799.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF160476.1 transcripts.
Set Membership: |
|

STAB2
Genomic Map