Assay Details
| Gene Symbol: LOC93463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135202.1 | 4 - 5 | 729 | 77 | ||
| GenBank mRNA | AL122100.1 | - | 4 - 5 | 729 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | atypical chemokine receptor 3 | ACKR3 |
CMKOR1, CXC-R7, CXCR-7, CXCR7, GPR159, RDC-1, RDC1 |
-468.6kb | Anti-sense | Chr. 2 - 236537131 - 236582358 on NCBI Build GRCh38 |
| Downstream | COP9 signalosome subunit 8 | COPS8 |
COP9, CSN8, SGN8 |
31.0kb | Anti-sense | Chr. 2 - 237085441 - 237098846 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC93463
Genomic Map