| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013286.hg.1 |
ENST00000302451.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013286.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16013286.hg.1 |
TC16000925.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
ENST00000575561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009656.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600173947.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600173947.hg.1 |
ENST00000575561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600173947.hg.1 |
NM_174924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600173947.hg.1 |
TC1600009656.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600173947.hg.1 |
ENST00000302451.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600173947.hg.1 |
ENST00000575561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600173947.hg.1 |
NM_174924.1 |
Refseq |