| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
ENST00000341272.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
ENST00000373808.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
ENST00000373818.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
ENST00000373823.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
ENST00000394353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
ENST00000412819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
ENST00000436847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
ENST00000449733.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
ENST00000456109.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
ENST00000545652.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
NM_000177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
NM_001127662.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
NM_001127663.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
NM_001127664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
NM_001127665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
NM_001127666.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
NM_001127667.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
NM_198252.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006948.hg.1 |
TC09000608.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
ENST00000341272.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
ENST00000373808.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
ENST00000373818.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
ENST00000373823.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
ENST00000394352.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
ENST00000394353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
ENST00000412819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
ENST00000436847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
ENST00000449733.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
ENST00000456109.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
ENST00000545652.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
NM_000177.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
NM_001127662.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
NM_001127663.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
NM_001127664.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
NM_001127665.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
NM_001127666.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
NM_001127667.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
NM_198252.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006947.hg.1 |
TC09000608.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000341272.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000373808.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000373818.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000394353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000412819.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000432226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000436847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000449733.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000449773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000475428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000477104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000477863.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000483960.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000545652.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_000177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127662.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127663.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127666.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127667.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001258029.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001258030.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_198252.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000341272.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000373808.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000373818.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000394353.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000412819.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000432226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000436847.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000449733.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000449773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000475428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000477104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000477863.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000483960.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
ENST00000545652.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_000177.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127662.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127663.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127664.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127665.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127666.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001127667.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001258029.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_001258030.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
NM_198252.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900184557.hg.1 |
TC0900012167.hg.1 |
Clariom D TC |