| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019194.hg.1 |
ENST00000349780.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019194.hg.1 |
ENST00000359309.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019194.hg.1 |
ENST00000360190.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019194.hg.1 |
ENST00000360822.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019194.hg.1 |
NM_001011649.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019194.hg.1 |
NM_018249.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09019194.hg.1 |
TC09001536.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000349780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000360190.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000416449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000473282.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000480112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000483412.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NM_001011649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NM_018249.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NR_073554.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NR_073555.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NR_073556.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NR_073557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NR_073558.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000349780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000360190.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000416449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000473282.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000480112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
ENST00000483412.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NM_001011649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NM_018249.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NR_073554.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NR_073555.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NR_073556.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NR_073557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
NR_073558.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900177479.hg.1 |
TC0900011378.hg.1 |
Clariom D TC |