Assay Details
| Gene Symbol: MYH15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014981.1 | NP_055796.1 | 37 - 38 | 5381 | 76 | |
| XM_011512559.2 | XP_011510861.1 | 38 - 39 | 5502 | 76 | ||
| XM_017005922.1 | XP_016861411.1 | 28 - 29 | 4646 | 76 | ||
| GenBank mRNA | AB023217.3 | - | 37 - 38 | 5381 | 76 | |
| AK055436.1 | - | 14 - 15 | 2467 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001631.hg.1
Set Membership: |
|

MYH15
Genomic Map