| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012334.hg.1 |
ENST00000317734.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012334.hg.1 |
NM_015945.11 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012334.hg.1 |
NM_173073.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012334.hg.1 |
NM_173179.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012334.hg.1 |
TC20000899.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012338.hg.1 |
NM_015945.11 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012338.hg.1 |
NM_173073.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012338.hg.1 |
NM_173179.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012338.hg.1 |
TC20000899.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012336.hg.1 |
ENST00000243896.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012336.hg.1 |
ENST00000317734.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012336.hg.1 |
ENST00000372223.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012336.hg.1 |
ENST00000543605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012336.hg.1 |
NM_015945.11 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012336.hg.1 |
NM_173073.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012336.hg.1 |
NM_173179.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012336.hg.1 |
TC20000899.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012337.hg.1 |
ENST00000243896.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012337.hg.1 |
ENST00000317734.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012337.hg.1 |
ENST00000543605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012337.hg.1 |
NM_015945.11 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012337.hg.1 |
NM_173073.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012337.hg.1 |
NM_173179.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012337.hg.1 |
TC20000899.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012333.hg.1 |
ENST00000243896.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012333.hg.1 |
ENST00000317734.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012333.hg.1 |
NM_015945.11 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012333.hg.1 |
NM_173073.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012333.hg.1 |
NM_173179.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012333.hg.1 |
TC20000899.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012335.hg.1 |
ENST00000317734.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012335.hg.1 |
NM_015945.11 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012335.hg.1 |
NM_173073.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012335.hg.1 |
NM_173179.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012335.hg.1 |
TC20000899.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012320.hg.1 |
ENST00000243896.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012320.hg.1 |
ENST00000372227.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012320.hg.1 |
NM_173179.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20012320.hg.1 |
TC20000899.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000164900.hg.1 |
ENST00000243896.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164900.hg.1 |
ENST00000372227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164900.hg.1 |
ENST00000484188.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164900.hg.1 |
NM_001281460.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164900.hg.1 |
NM_173179.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164900.hg.1 |
TC2000009267.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164900.hg.1 |
ENST00000243896.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164900.hg.1 |
ENST00000372227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164900.hg.1 |
ENST00000484188.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164900.hg.1 |
NM_001281460.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164900.hg.1 |
NM_173179.3 |
Refseq |