Assay Details
| Gene Symbol: UIMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005265932.2 | XP_005265989.1 | 4 - 5 | 478 | 87 | |
| XM_005265933.2 | XP_005265990.1 | 4 - 5 | 590 | 87 | ||
| XM_017009576.1 | XP_016865065.1 | 4 - 5 | 491 | 87 | ||
| XM_017009577.1 | XP_016865066.1 | 4 - 5 | 543 | 87 | ||
More Information
Set Membership: |
|

UIMC1
Genomic Map