Assay Details
| Gene Symbol: MTERF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301135.1 | NP_001288064.1 | 1 - 2 | 64 | 94 | |
| NM_006980.4 | NP_008911.1 | 1 - 2 | 64 | 94 | ||
| GenBank mRNA | AK222730.1 | - | 1 - 2 | 64 | 94 | |
| AK313641.1 | - | 1 - 2 | 46 | 94 | ||
| Y09615.1 | - | 1 - 2 | 47 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001595.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK313641.1,NM_001301135.1,NM_006980.4,Y09615.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07024085.hg.1 | ENST00000351870.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024085.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024085.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024085.hg.1 | TC07001595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024086.hg.1 | ENST00000351870.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024086.hg.1 | ENST00000406735.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024086.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024086.hg.1 | ENST00000454222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024086.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024086.hg.1 | TC07001595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024087.hg.1 | ENST00000351870.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024087.hg.1 | ENST00000406735.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024087.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024087.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024087.hg.1 | TC07001595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024071.hg.1 | ENST00000351870.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024071.hg.1 | ENST00000406735.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024071.hg.1 | ENST00000454222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024071.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024071.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07024071.hg.1 | TC07001595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000442961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000442961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000442961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000442961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000442961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000442961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201092.hg.1 | TC0700013583.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000351870.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000419292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000425936.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000442961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000442961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000442961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000442961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301134.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013583.hg.1 | NM_006980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201090.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201090.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201090.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201090.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201090.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201090.hg.1 | ENST00000456229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201090.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201090.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201090.hg.1 | TC0700013583.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700201091.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201091.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201091.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201091.hg.1 | ENST00000406735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201091.hg.1 | ENST00000481516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201091.hg.1 | NM_001301135.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201091.hg.1 | TC0700013583.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 102116 | - | - |

MTERF1
Genomic Map