Assay Details
| Gene Symbol: EDNRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045958.1 | 3 - 4 | 1078 | 63 | ||
| GenBank mRNA | S81542.1 | - | 2 - 3 | 548 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EDNRA| Gene Symbol: EDNRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045958.1 | 3 - 4 | 1078 | 63 | ||
| GenBank mRNA | S81542.1 | - | 2 - 3 | 548 | 63 | |
Set Membership: |
|